Assay Details
Target Gene Details
Entrez Gene ID: | 75540 |
Gene Name: | fucose-1-phosphate guanylyltransferase |
Gene Aliases: |
1700016E03Rik |
Location: |
Chr.3:155084915-155093378 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fpgt | NM_029330.2 | 2 | 122 | NP_083606.2 |
| AJ276067.2 | 2 | 109 | CAC81971.2 | |
| AK028640.1 | 2 | 119 | BAC26043.1 | |
| AK045369.1 | 2 | 123 | ||
| BC110551.1 | 2 | 122 | ||
| BC110552.1 | 2 | 119 | AAI10553.1 |
More Information
Set Membership: |
|

Genomic Map