Assay Details
Target Gene Details
Entrez Gene ID: | 229900 |
Gene Name: | guanylate binding protein 7 |
Gene Aliases: |
9830147J24Rik |
Location: |
Chr.3:142530336-142550151 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbp7 | NM_001083312.2 | 2 | 205 | NP_001076781.1 |
| NM_145545.4 | 2 | 223 | NP_663520.2 | |
| AK036666.1 | 2 | 219 | BAC29526.1 | |
| AK087751.1 | 2 | 199 | BAC39989.1 | |
| AK131176.1 | 1 | 106 | BAD21426.1 | |
| BC010229.1 | 2 | 170 | ||
| BY742706.1 | 2 | 203 |
More Information
Set Membership: |
|

Genomic Map