Assay Details
Target Gene Details
Entrez Gene ID: | 14800 |
Gene Name: | glutamate receptor, ionotropic, AMPA2 (alpha 2) |
Gene Aliases: |
GluA2, GluR-B, Glur-2, Glur2 |
Location: |
Chr.3:80684935-80803204 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gria2 | NM_001039195.1 | NP_001034284.1 | ||
| NM_001083806.1 | NP_001077275.1 | |||
| NM_013540.2 | NP_038568.2 | |||
| AB109210.1 | ||||
| AB109211.1 | ||||
| AB111957.1 | BAE06155.1 | |||
| AK014389.1 | BAB29316.1 | |||
| AK043490.1 | BAC31557.1 | |||
| AK044574.1 | BAC31986.1 | |||
| AK046861.1 | BAC32899.1 | |||
| AK086614.1 | ||||
| AK134572.1 | ||||
| AK134647.1 | ||||
| AK134772.1 | ||||
| AK141525.1 | ||||
| AK163941.1 | ||||
| BC048248.1 | AAH48248.1 | |||
| X57498.1 | CAA40735.1 |
More Information
Set Membership: |
|

Genomic Map