Assay Details
Target Gene Details
Entrez Gene ID: | 67553 |
Gene Name: | glutathione S-transferase, C-terminal domain containing |
Gene Aliases: |
4933434L15Rik |
Location: |
Chr.3:132981752-133091971 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstcd | AK029724.1 | 1 | 118 | |
| BC061240.1 | 1 | 115 | AAH61240.1 |

Genomic Map