Assay Details
Target Gene Details
Entrez Gene ID: | 15360 |
Gene Name: | 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 |
Gene Aliases: |
1300002P16, mHS |
Location: |
Chr.3:98280431-98310738 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgcs2 | NM_008256.4 | 3 | 674 | NP_032282.2 |
| AK004865.1 | 3 | 672 | BAB23626.1 | |
| AK004902.1 | 3 | 670 | BAB23657.1 | |
| AK098104.1 | 3 | 670 | BAC05233.1 | |
| BC014714.1 | 3 | 660 | AAH14714.1 | |
| BC024744.1 | 3 | 658 | AAH24744.1 |

Genomic Map