Assay Details
Target Gene Details
Entrez Gene ID: | 30942 |
Gene Name: | hepatocyte nuclear factor 4, gamma |
Gene Aliases: |
NR2A2 |
Location: |
Chr.3:3508030-3659800 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnf4g | NM_013920.2 | 2 | 386 | NP_038948.1 |
| AJ242626.1 | 2 | 338 | CAB43724.1 | |
| AK143816.1 | 2 | 387 | BAE25549.1 | |
| BC125519.1 | 2 | 208 |
More Information
Set Membership: |
|

Genomic Map