Assay Details
Target Gene Details
Entrez Gene ID: | 66059 |
Gene Name: | keratinocyte associated protein 2 |
Gene Aliases: |
0610010I12Rik, Kcp2 |
Location: |
Chr.3:89246300-89249723 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtcap2 | NM_025327.2 | NP_079603.1 | ||
| AK002479.1 | BAB22132.1 | |||
| AK003310.1 | BAB22707.1 | |||
| BC031492.1 | AAH31492.1 | |||
| BC043030.1 | AAH43030.1 |

Genomic Map