Assay Details
Target Gene Details
Entrez Gene ID: | 12428 |
Gene Name: | cyclin A2 |
Gene Aliases: |
AA408589, Ccn-1, Ccn1, Ccna, CycA2, Cyca |
Location: |
Chr.3:36564865-36571996 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccna2 | NM_009828.2 | 2 | 520 | NP_033958.2 |
| AK044924.1 | 2 | 523 | BAC32144.1 | |
| AK135861.1 | 2 | 492 | ||
| AK188335.1 | 2 | 187 | ||
| AK216847.1 | 2 | 192 | ||
| BB642910.1 | 2 | 520 | ||
| BC052730.1 | 2 | 468 | AAH52730.1 | |
| X75483.1 | 2 | 468 | CAA53212.1 | |
| Z26580.1 | 2 | 477 | CAA81331.1 |
More Information
Set Membership: |
|

Genomic Map