Assay Details
Target Gene Details
Entrez Gene ID: | 12428 |
Gene Name: | cyclin A2 |
Gene Aliases: |
AA408589, Ccn-1, Ccn1, Ccna, CycA2, Cyca |
Location: |
Chr.3:36564865-36571996 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccna2 | NM_009828.2 | 4 | 870 | NP_033958.2 |
| AK044924.1 | 4 | 873 | BAC32144.1 | |
| AK135861.1 | 4 | 842 | ||
| AK180714.1 | 2 | 177 | ||
| AK182507.1 | 2 | 177 | ||
| AK190337.1 | 2 | 177 | ||
| AK192533.1 | 2 | 177 | ||
| AK203680.1 | 2 | 177 | ||
| AK215356.1 | 2 | 177 | ||
| BC052730.1 | 4 | 818 | AAH52730.1 | |
| X75483.1 | 4 | 818 | CAA53212.1 | |
| Z26580.1 | 4 | 827 | CAA81331.1 |
More Information
Set Membership: |
|

Genomic Map