Assay Details
Target Gene Details
Entrez Gene ID: | 76915 |
Gene Name: | meiotic nuclear divisions 1 homolog (S. cerevisiae) |
Gene Aliases: |
2610034E18Rik, AI591601, Gaj |
Location: |
Chr.3:84087933-84156861 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mnd1 | NM_029797.3 | NP_084073.2 | ||
| AK011664.1 | BAB27765.1 | |||
| BC027741.1 | AAH27741.1 | |||
| BG070803.2 | ||||
| BY710901.1 |

Genomic Map