Assay Details
Target Gene Details
Entrez Gene ID: | 246691 |
Gene Name: | prokineticin 1 |
Gene Aliases: |
EG-VEGF |
Location: |
Chr.3:107231916-107239707 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prok1 | NM_001044382.1 | NP_001037847.1 | ||
| AF487281.1 | AAM49573.1 | |||
| AK044344.1 | 1 | 64 | ||
| BC042707.1 | ||||
| BC117006.1 | ||||
| BC117008.1 |
More Information
Set Membership: |
|

Genomic Map