Assay Details
Target Gene Details
Entrez Gene ID: | 66921 |
Gene Name: | PRP38 pre-mRNA processing factor 38 (yeast) domain containing B |
Gene Aliases: |
1110021E09Rik, AU018955 |
Location: |
Chr.3:108902807-108911752 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf38b | NM_025845.2 | NP_080121.1 | ||
| AK015938.1 | BAB30042.1 | |||
| AK018126.1 | BAB31082.1 | |||
| AK040788.1 | ||||
| AK159267.1 | ||||
| AK170138.1 | 1 | 2227 | ||
| BC050900.1 | AAH50900.1 | |||
| BC052077.1 | AAH52077.1 |

Genomic Map