Assay Details
Target Gene Details
Entrez Gene ID: | 67860 |
Gene Name: | S100 calcium binding protein A16 |
Gene Aliases: |
2300002L21Rik, AI325039, AI663996, DT1P1A7, S100F |
Location: |
Chr.3:90537257-90543151 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a16 | NM_026416.2 | 2 | 223 | NP_080692.1 |
| AK009762.1 | 2 | 174 | BAB26486.1 | |
| AK160178.1 | 2 | 174 | BAE35676.1 | |
| AK160512.1 | 2 | 166 | BAE35835.1 | |
| AK167006.1 | 2 | 166 | BAE39183.1 | |
| AY221962.1 | 1 | 114 | AAP46153.1 | |
| BC020031.1 | 2 | 223 | AAH20031.1 |
More Information
Set Membership: |
|

Genomic Map