Assay Details
Target Gene Details
Entrez Gene ID: | 56758 |
Gene Name: | muscleblind-like 1 (Drosophila) |
Gene Aliases: |
Mbnl, mKIAA0428 |
Location: |
Chr.3:60472791-60629750 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbnl1 | NM_001253708.1 | 4 | 1492 | NP_001240637.1 |
| NM_001253709.1 | 3 | 443 | NP_001240638.1 | |
| NM_001253710.1 | 3 | 443 | NP_001240639.1 | |
| NM_001253711.1 | 3 | 443 | NP_001240640.1 | |
| NM_001253713.1 | 4 | 446 | NP_001240642.1 | |
| NM_020007.3 | 4 | 1492 | NP_064391.2 | |
| AB093240.1 | 4 | 1496 | BAC41424.1 | |
| AF231110.1 | 3 | 932 | AAF72159.1 | |
| AK038242.1 | 3 | 527 | BAC29950.1 | |
| AK149405.1 | ||||
| AK153827.1 | 3 | 336 | BAE32199.1 | |
| AK153848.1 | 3 | 431 | BAE32210.1 | |
| BC096600.1 | 3 | 404 | AAH96600.1 | |
| CN663545.1 |
More Information
Set Membership: |
|

Genomic Map