Assay Details
Target Gene Details
Entrez Gene ID: | 56758 |
Gene Name: | muscleblind-like 1 (Drosophila) |
Gene Aliases: |
Mbnl, mKIAA0428 |
Location: |
Chr.3:60472791-60629750 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbnl1 | NM_001253708.1 | 1 | 38 | NP_001240637.1 |
| NM_001253709.1 | 1 | 38 | NP_001240638.1 | |
| NM_001253710.1 | 1 | 38 | NP_001240639.1 | |
| NM_001253711.1 | 1 | 38 | NP_001240640.1 | |
| NM_001253713.1 | NP_001240642.1 | |||
| NM_020007.3 | 1 | 38 | NP_064391.2 | |
| AB093240.1 | 1 | 38 | BAC41424.1 | |
| AK088871.1 | ||||
| AK149405.1 | 1 | 54 | ||
| AK153827.1 | BAE32199.1 | |||
| AK153848.1 | BAE32210.1 | |||
| AK162039.1 | 1 | 71 | ||
| BB841817.1 | ||||
| BC096600.1 | AAH96600.1 |
More Information
Set Membership: |
|

Genomic Map