Assay Details
Target Gene Details
Entrez Gene ID: | 67709 |
Gene Name: | regenerating islet-derived family, member 4 |
Gene Aliases: |
2010002L15Rik, GISP, RELP |
Location: |
Chr.3:98222156-98236748 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reg4 | NM_026328.2 | 2 | 183 | NP_080604.2 |
| AK008049.1 | 2 | 185 | BAB25429.1 | |
| AK008438.1 | 2 | 186 | BAB25669.1 | |
| AK160329.1 | 2 | 180 | ||
| AK162316.1 | 2 | 184 | BAE36851.1 | |
| BC019465.1 | 2 | 166 | AAH19465.1 |

Genomic Map