Assay Details
Target Gene Details
Entrez Gene ID: | 67709 |
Gene Name: | regenerating islet-derived family, member 4 |
Gene Aliases: |
2010002L15Rik, GISP, RELP |
Location: |
Chr.3:98222156-98236748 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reg4 | NM_026328.2 | NP_080604.2 | ||
| AK008049.1 | BAB25429.1 | |||
| AK008438.1 | BAB25669.1 | |||
| AK160329.1 | ||||
| AK162316.1 | BAE36851.1 | |||
| BC019465.1 | AAH19465.1 |

Genomic Map