Assay Details
Target Gene Details
Entrez Gene ID: | 20199 |
Gene Name: | S100 calcium binding protein A5 |
Gene Aliases: |
S100D9 |
Location: |
Chr.3:90606479-90611784 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a5 |

Genomic Map