Assay Details
Target Gene Details
Entrez Gene ID: | 20199 |
Gene Name: | S100 calcium binding protein A5 |
Gene Aliases: |
S100D9 |
Location: |
Chr.3:90606479-90611784 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a5 | NM_011312.2 | 1 | 87 | NP_035442.1 |
| AF087469.1 | 1 | 87 | AAC64106.1 | |
| AI327447.1 | 1 | 87 | ||
| BC117108.1 | 1 | 86 |
More Information
Set Membership: |
|

Genomic Map