Assay Details
Target Gene Details
Entrez Gene ID: | 20200 |
Gene Name: | S100 calcium binding protein A6 (calcyclin) |
Gene Aliases: |
2A9, 5B10, CALCYCLIN, Cacy, PRA |
Location: |
Chr.3:90612894-90614414 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a6 | NM_011313.2 | 2 | 456 | NP_035443.1 |
| AF549497.1 | 2 | 164 | AAN59899.1 | |
| AK008241.1 | 2 | 167 | BAB25552.1 | |
| AK008941.1 | 2 | 167 | BAB25982.1 | |
| AK150408.1 | 2 | 167 | BAE29534.1 | |
| AK151144.1 | 2 | 167 | BAE30151.1 | |
| AK151587.1 | 2 | 167 | BAE30527.1 | |
| BC003832.1 | 2 | 219 | AAH03832.1 | |
| BC010774.1 | 2 | 181 | AAH10774.1 | |
| BM932500.1 | 2 | 468 | ||
| M37761.1 | 2 | 165 | AAA37358.1 | |
| X52278.1 | 2 | 177 | CAA36521.1 |
More Information
Set Membership: |
|

Genomic Map