Assay Details
Target Gene Details
Entrez Gene ID: | 72039 |
Gene Name: | methylcrotonoyl-Coenzyme A carboxylase 1 (alpha) |
Gene Aliases: |
1810045E08Rik, 2310058B18Rik, MCCalpha, Mcca, R75106 |
Location: |
Chr.3:35959296-36000678 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mccc1 | NM_023644.4 | 17 | 2032 | NP_076133.3 |
| AF310338.1 | 17 | 1953 | AAG50244.1 | |
| AK007782.1 | 17 | 1985 | BAB25253.1 | |
| AK031072.1 | 17 | 1993 | BAC27239.1 | |
| AK032203.1 | 17 | 1991 | ||
| AK141040.1 | 2 | 4840 | ||
| AK168589.1 | 17 | 1921 | BAE40458.1 | |
| BC021382.1 | 17 | 1996 | AAH21382.1 |
More Information
Set Membership: |
|

Genomic Map