Assay Details
Target Gene Details
Entrez Gene ID: | 53970 |
Gene Name: | regulatory factor X, 5 (influences HLA class II expression) |
Gene Aliases: |
- |
Location: |
Chr.3:94951957-94961561 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfx5 | NM_017395.2 | NP_059091.2 | ||
| AB221679.1 | ||||
| AF209854.1 | AAF68260.1 | |||
| AK088686.1 | 6 | 567 | ||
| BB666132.1 | ||||
| BC051965.1 | ||||
| BC075717.1 | AAH75717.1 | |||
| BC118948.2 | ||||
| BC118949.2 |
More Information
Set Membership: |
|

Genomic Map