Assay Details
Target Gene Details
Entrez Gene ID: | 72039 |
Gene Name: | methylcrotonoyl-Coenzyme A carboxylase 1 (alpha) |
Gene Aliases: |
1810045E08Rik, 2310058B18Rik, MCCalpha, Mcca, R75106 |
Location: |
Chr.3:35959296-36000678 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mccc1 | NM_023644.4 | NP_076133.3 | ||
| AF310338.1 | AAG50244.1 | |||
| AK007782.1 | BAB25253.1 | |||
| AK031072.1 | BAC27239.1 | |||
| AK032203.1 | ||||
| AK160184.1 | BAE35680.1 | |||
| AK168589.1 | BAE40458.1 | |||
| BC021382.1 | AAH21382.1 |
More Information
Set Membership: |
|

Genomic Map