Assay Details
Target Gene Details
Entrez Gene ID: | 241915 |
Gene Name: | polyhomeotic-like 3 (Drosophila) |
Gene Aliases: |
E030046K01Rik, Edr3, Hph3 |
Location: |
Chr.3:30899295-30969479 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phc3 | NM_001165954.1 | NP_001159426.1 | ||
| NM_001165955.1 | NP_001159427.1 | |||
| NM_001165956.1 | NP_001159428.1 | |||
| NM_153421.2 | NP_700470.2 | |||
| AJ414610.1 | CAC93885.1 | |||
| AK037348.1 | ||||
| AK051918.1 | 1 | 1256 | ||
| AK087339.1 | ||||
| AK166208.1 | ||||
| AK166296.1 | BAE38687.1 | |||
| BC132533.1 | ||||
| BC145123.1 |
More Information
Set Membership: |
|

Genomic Map