Assay Details
Target Gene Details
Entrez Gene ID: | 19185 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 4 |
Gene Aliases: |
Af1, Mcb1, angiocidin |
Location: |
Chr.3:95032688-95042614 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd4 | NM_001282017.1 | NP_001268946.1 | ||
| NM_008951.2 | NP_032977.1 | |||
| AB029142.1 | BAA97573.1 | |||
| AB029143.1 | BAA97574.1 | |||
| AB029144.1 | 9 | 1031 | BAA97575.1 | |
| AB029145.1 | 8 | 1110 | BAA97576.1 | |
| AF013099.1 | AAC53547.1 | |||
| AF175574.1 | AAG09199.1 | |||
| AK010969.1 | BAB27299.1 | |||
| AK145524.1 | BAE26484.1 | |||
| BC009005.1 | AAH09005.1 | |||
| BY473933.1 |
More Information
Set Membership: |
|

Genomic Map