Assay Details
Target Gene Details
Entrez Gene ID: | 19769 |
Gene Name: | Ras-like without CAAX 1 |
Gene Aliases: |
RIBB, ROC1, Rit |
Location: |
Chr.3:88716840-88731048 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rit1 | NM_001163310.1 | NP_001156782.1 | ||
| NM_009069.4 | NP_033095.1 | |||
| NR_028056.1 | ||||
| AK005357.1 | BAB23972.1 | |||
| AK014126.1 | BAB29168.1 | |||
| AK018785.1 | 4 | 377 | ||
| AK164196.1 | ||||
| AK168254.1 | 4 | 388 | ||
| AK184013.1 | ||||
| BC012694.1 | AAH12694.1 | |||
| BY284832.1 | ||||
| CV562852.1 | ||||
| CX242151.1 | ||||
| U71205.1 | AAB42215.1 |
More Information
Set Membership: |
|

Genomic Map