Assay Details
Target Gene Details
Entrez Gene ID: | 74442 |
Gene Name: | sphingomyelin synthase 2 |
Gene Aliases: |
4933405A16Rik, 5133401H06Rik, AI854299 |
Location: |
Chr.3:131318985-131491192 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgms2 | NM_028943.5 | NP_083219.2 | ||
| AK016659.1 | BAB30364.2 | |||
| BC117782.2 |

Genomic Map