Assay Details
Target Gene Details
Entrez Gene ID: | 66256 |
Gene Name: | signal sequence receptor, beta |
Gene Aliases: |
1500032E05Rik, AI315033, AU020133, TLAP, TRAPB, TRAPbeta |
Location: |
Chr.3:88579584-88588420 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssr2 | NM_025448.3 | NP_079724.1 | ||
| AK005340.1 | BAB23962.1 | |||
| AK007538.1 | BAB25097.1 | |||
| AK007720.1 | BAB25211.1 | |||
| AK010565.1 | BAB27030.1 | |||
| AK012281.1 | ||||
| AK020145.1 | BAB32009.1 | |||
| AK050505.1 | BAC34295.1 | |||
| AK084530.1 | BAC39212.1 | |||
| AK213548.1 | ||||
| AK214352.1 | ||||
| BC010214.1 | AAH10214.1 |
More Information
Set Membership: |
|

Genomic Map