Assay Details
Target Gene Details
Entrez Gene ID: | 229615 |
Gene Name: | protein inhibitor of activated STAT 3 |
Gene Aliases: |
Pias3l |
Location: |
Chr.3:96696375-96706070 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pias3 | NM_001165949.1 | 2 | 281 | NP_001159421.1 |
| NM_018812.2 | 2 | 281 | NP_061282.2 | |
| NM_146135.2 | 2 | 318 | NP_666247.1 | |
| AF034080.3 | 1 | 158 | ||
| AK040619.1 | 2 | 309 | BAC30647.1 | |
| AK077551.1 | 2 | 267 | BAC36858.1 | |
| AK085503.1 | 2 | 252 | ||
| AK211740.1 | ||||
| BC023128.1 | 2 | 224 | AAH23128.1 | |
| BC051252.1 | 2 | 191 | AAH51252.1 | |
| BY002850.1 | 2 | 323 | ||
| BY154092.1 | 2 | 329 |

Genomic Map