Assay Details
Target Gene Details
Entrez Gene ID: | 17235 |
Gene Name: | sperm mitochondria-associated cysteine-rich protein |
Gene Aliases: |
Mcs, Mcsp |
Location: |
Chr.3:92583866-92589024 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smcp | NM_008574.3 | 1 | 144 | NP_032600.2 |
| AK077172.1 | 1 | 115 | BAE43359.1 | |
| BC055104.1 | 1 | 114 | AAH55104.2 | |
| BC061167.1 | 1 | 94 | AAH61167.1 | |
| BU936528.1 | 1 | 147 | ||
| M29603.1 | 1 | 117 | AAA53045.3 | |
| S49659.1 | 1 | 114 | AAC08283.1 |

Genomic Map