Assay Details
Target Gene Details
Entrez Gene ID: | 67437 |
Gene Name: | signal sequence receptor, gamma |
Gene Aliases: |
0610038P07Rik, AL022999, AU022074, AW553833, TRAPG |
Location: |
Chr.3:65379657-65392553 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssr3 | NM_026155.3 | NP_080431.1 | ||
| AK002818.1 | BAB22381.1 | |||
| AK029353.1 | BAC26413.1 | |||
| AK076235.1 | BAC36268.1 | |||
| AK088555.1 | BAC40420.1 | |||
| AK150255.1 | BAE29416.1 | |||
| AK162235.1 | BAE36806.1 | |||
| BC011111.1 | AAH11111.1 | |||
| CT010172.1 | CAJ18380.1 |

Genomic Map