Assay Details
Target Gene Details
Entrez Gene ID: | 229302 |
Gene Name: | transmembrane 4 superfamily member 4 |
Gene Aliases: |
Iltmp |
Location: |
Chr.3:57425410-57441677 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm4sf4 | NM_145539.2 | NP_663514.2 | ||
| AK148484.1 | BAE28579.1 | |||
| AK165731.1 | BAE38358.1 | |||
| BC010814.1 | AAH10814.1 |

Genomic Map