Assay Details
Target Gene Details
Entrez Gene ID: | 109245 |
Gene Name: | leucine rich repeat containing 39 |
Gene Aliases: |
2010005E21Rik, 9430028I06Rik |
Location: |
Chr.3:116562953-116583134 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc39 | NM_027321.3 | NP_081597.1 | ||
| NM_175413.3 | NP_780622.1 | |||
| AK008119.1 | BAB25474.1 | |||
| AK035072.1 | BAC28932.1 | |||
| AK039277.1 | BAC30303.1 | |||
| AK046671.1 | BAC32831.1 | |||
| AK052159.1 | BAC34863.1 | |||
| AK079125.1 | BAC37551.1 | |||
| BC119529.2 | ||||
| BC119530.1 |
More Information
Set Membership: |
|

Genomic Map