Assay Details
Target Gene Details
Entrez Gene ID: | 75624 |
Gene Name: | methionyl aminopeptidase 1 |
Gene Aliases: |
1700029C17Rik, AW047992, mKIAA0094 |
Location: |
Chr.3:138458960-138489382 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Metap1 | NM_175224.4 | NP_780433.1 | ||
| AK006484.1 | ||||
| AK077694.1 | BAC36961.1 | |||
| AK129054.1 | BAC97864.2 | |||
| AK152138.1 | BAE30977.1 | |||
| AK153231.1 | BAE31824.1 | |||
| AK153944.1 | ||||
| BC027110.1 | AAH27110.1 | |||
| BC096469.1 | AAH96469.1 |
More Information
Set Membership: |
|

Genomic Map