Assay Details
Target Gene Details
Entrez Gene ID: | 229615 |
Gene Name: | protein inhibitor of activated STAT 3 |
Gene Aliases: |
Pias3l |
Location: |
Chr.3:96696375-96706070 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pias3 | NM_146135.2 | NP_666247.1 | ||
| AK040619.1 | BAC30647.1 | |||
| AK077551.1 | BAC36858.1 | |||
| AK085503.1 | ||||
| BC023128.1 | AAH23128.1 | |||
| BY002850.1 |

Genomic Map