Assay Details
Target Gene Details
Entrez Gene ID: | 67161 |
Gene Name: | sodium channel and clathrin linker 1 |
Gene Aliases: |
2610207F23Rik, 4931421F20Rik |
Location: |
Chr.3:41626705-41742530 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sclt1 | NM_001081411.1 | NP_001074880.1 | ||
| AK011907.1 | BAB27908.1 | |||
| AK014777.1 | BAB29547.1 | |||
| AK016469.1 | BAC25482.2 | |||
| AK083894.1 | ||||
| BC026498.1 | AAH26498.1 | |||
| BC043136.1 | AAH43136.1 | |||
| BC060271.1 | AAH60271.1 |

Genomic Map