Assay Details
Target Gene Details
Entrez Gene ID: | 210529 |
Gene Name: | methyltransferase like 14 |
Gene Aliases: |
G430022H21Rik, mKIAA1627 |
Location: |
Chr.3:123368295-123385990 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl14 | NM_201638.2 | 9 | 975 | NP_964000.2 |
| AK089949.1 | 9 | 922 | ||
| AK146881.1 | 9 | 964 | BAE27502.1 | |
| AK153810.1 | 9 | 971 | ||
| BC052204.1 | 9 | 933 | AAH52204.1 |
More Information
Set Membership: |
|

Genomic Map