Assay Details
Target Gene Details
Entrez Gene ID: | 19729 |
Gene Name: | solute carrier family 50 (sugar transporter), member 1 |
Gene Aliases: |
MmSWEET1, Rag1ap1, Rga |
Location: |
Chr.3:89268246-89270570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc50a1 | NM_009057.3 | NP_033083.2 | ||
| AK014305.1 | BAB29259.1 | |||
| AK045924.1 | 1 | 476 | ||
| AK079716.1 | 2 | 228 | ||
| AK158862.1 | 2 | 188 | ||
| AK170162.1 | BAE41608.1 | |||
| AK177059.1 | ||||
| BC014292.1 | AAH14292.1 | |||
| BY333933.1 | ||||
| X96618.1 | CAA65438.1 |

Genomic Map