Assay Details
Target Gene Details
Entrez Gene ID: | 99683 |
Gene Name: | Sec24 related gene family, member B (S. cerevisiae) |
Gene Aliases: |
AI605202, SEC24 |
Location: |
Chr.3:129982764-130061573 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec24b | NM_207209.2 | 14 | 2590 | NP_997092.1 |
| AK150076.1 | 13 | 1667 | BAE29287.1 | |
| AK152339.1 | 12 | 1562 | BAE31136.1 | |
| BC046776.1 | 14 | 2590 | AAH46776.1 | |
| BC057355.1 | 14 | 2451 | AAH57355.1 |
More Information
Set Membership: |
|

Genomic Map