Assay Details
Target Gene Details
Entrez Gene ID: | 68436 |
Gene Name: | ribosomal protein L34 |
Gene Aliases: |
1100001I22Rik |
Location: |
Chr.3:130726827-130730398 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl34 | NM_001005859.3 | NP_001005859.1 | ||
| NM_001287581.1 | NP_001274510.1 | |||
| NM_026724.2 | NP_081000.1 | |||
| AK003177.1 | BAB22621.1 | |||
| AK146252.1 | BAE27014.1 | |||
| BC028517.1 | AAH28517.1 | |||
| BC058118.1 | AAH58118.1 | |||
| CJ041925.1 | ||||
| CJ043938.1 | ||||
| CJ044447.1 |
More Information
Set Membership: |
|

Genomic Map