Assay Details
Target Gene Details
Entrez Gene ID: | 50874 |
Gene Name: | tropomodulin 4 |
Gene Aliases: |
MTMOD, Sk-Tmod |
Location: |
Chr.3:95124117-95129208 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmod4 | NM_016712.3 | NP_057921.1 | ||
| AF177174.1 | AAF31673.1 | |||
| AK144408.1 | BAE25875.1 | |||
| AK144535.1 | BAE25929.1 | |||
| BC068020.1 | AAH68020.1 |
More Information
Set Membership: |
|

Genomic Map