Assay Details
Target Gene Details
Entrez Gene ID: | 57294 |
Gene Name: | ribosomal protein S27 |
Gene Aliases: |
3200001M24Rik |
Location: |
Chr.3:90212667-90213648 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps27 | NM_027015.4 | NP_081291.1 | ||
| NR_033727.1 | ||||
| AA036204.1 | ||||
| AK014294.1 | BAB29250.1 | |||
| AK084058.1 | ||||
| AK088320.1 | BAC40279.1 | |||
| AK166551.1 | BAE38849.1 | |||
| BC048352.1 | AAH48352.1 | |||
| BC055693.1 | AAH55693.1 |
More Information
Set Membership: |
|

Genomic Map