Assay Details
Target Gene Details
Entrez Gene ID: | 69608 |
Gene Name: | Sec24 related gene family, member D (S. cerevisiae) |
Gene Aliases: |
2310020L09Rik, Gm1349 |
Location: |
Chr.3:123267496-123365641 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec24d | NM_027135.2 | 17 | 2283 | NP_081411.2 |
| BC026624.1 | 4 | 407 | AAH26624.1 | |
| BC049257.1 | 6 | 676 | AAH49257.1 | |
| BC067020.1 | 17 | 2283 | AAH67020.1 |
More Information
Set Membership: |
|

Genomic Map