Assay Details
Target Gene Details
Entrez Gene ID: | 20526 |
Gene Name: | solute carrier family 2 (facilitated glucose transporter), member 2 |
Gene Aliases: |
AI266973, Glut-2, Glut2 |
Location: |
Chr.3:28697903-28728361 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc2a2 | NM_031197.2 | 10 | 1424 | NP_112474.2 |
| AK005068.1 | 10 | 1425 | BAB23792.1 | |
| AK149532.1 | 3 | 537 | ||
| AK165430.1 | 10 | 1359 | BAE38184.1 | |
| BC034675.1 | 10 | 1343 | AAH34675.1 | |
| S77926.1 | 2 | 163 | AAB20847.1 | |
| X15684.1 | 10 | 1373 | CAA33719.1 | |
| X16986.1 | 10 | 1363 | CAA34855.1 |

Genomic Map