Assay Details
Target Gene Details
Entrez Gene ID: | 69276 |
Gene Name: | SEC62 homolog (S. cerevisiae) |
Gene Aliases: |
3100002M17Rik, AI844545, AW545092, Dtrp1, HTP1, Tloc1 |
Location: |
Chr.3:30792774-30821263 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec62 | NM_027016.2 | NP_081292.1 | ||
| AK013922.1 | BAB29058.1 | |||
| AK088093.1 | BAC40140.1 | |||
| AK090078.1 | ||||
| BC017162.1 | ||||
| BC067202.1 | AAH67202.1 |

Genomic Map