Assay Details
Target Gene Details
Entrez Gene ID: | 69276 |
Gene Name: | SEC62 homolog (S. cerevisiae) |
Gene Aliases: |
3100002M17Rik, AI844545, AW545092, Dtrp1, HTP1, Tloc1 |
Location: |
Chr.3:30792774-30821263 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec62 | NM_027016.2 | NP_081292.1 | ||
| AK088093.1 | BAC40140.1 | |||
| AK090078.1 | ||||
| BC023720.1 | AAH23720.1 | |||
| BC067202.1 | AAH67202.1 |
More Information
Set Membership: |
|

Genomic Map