Assay Details
Target Gene Details
Entrez Gene ID: | 229877 |
Gene Name: | RAP1, GTP-GDP dissociation stimulator 1 |
Gene Aliases: |
BC011279, GDS1 |
Location: |
Chr.3:138925897-139075201 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rap1gds1 | NM_001040690.2 | NP_001035780.1 | ||
| NM_001286759.1 | NP_001273688.1 | |||
| NM_145544.3 | NP_663519.2 | |||
| AK052125.1 | BAC34852.1 | |||
| AK154852.1 | ||||
| AK155793.1 | ||||
| AK160989.1 | BAE36135.1 | |||
| AK164159.1 | BAE37656.1 | |||
| AK166315.1 | BAE38698.1 | |||
| AK172052.1 | BAE42801.1 | |||
| CF731381.1 | ||||
| CN702379.1 |

Genomic Map