Assay Details
Target Gene Details
Entrez Gene ID: | 11416 |
Gene Name: | solute carrier family 33 (acetyl-CoA transporter), member 1 |
Gene Aliases: |
AI315656, AI788741, Acatn, D630022N01Rik |
Location: |
Chr.3:63942323-63964733 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc33a1 | NM_001272035.1 | 3 | 1564 | NP_001258964.1 |
| NM_015728.5 | 3 | 1564 | NP_056543.2 | |
| AB016795.1 | 3 | 1021 | BAA76933.1 | |
| AK144703.1 | 3 | 1431 | ||
| AW519598.1 | 1 | 43 | ||
| BC005616.1 | 3 | 1141 | AAH05616.1 | |
| BC054395.1 | 3 | 1564 | AAH54395.1 |
More Information
Set Membership: |
|

Genomic Map