Assay Details
Target Gene Details
Entrez Gene ID: | 11610 |
Gene Name: | angiotensin II, type I receptor-associated protein |
Gene Aliases: |
3300002E14Rik, AT1R, Atrap, D4Wsu124e |
Location: |
Chr.4:148077061-148088064 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agtrap | NM_001301281.1 | 4 | 379 | NP_001288210.1 |
| NM_009642.4 | 4 | 426 | NP_033772.2 | |
| AF102548.1 | 4 | 305 | AAD25997.1 | |
| AK007383.1 | 4 | 363 | BAB25001.1 | |
| AK007502.1 | 4 | 338 | BAB25074.1 | |
| AK036598.1 | 4 | 355 | BAC29501.1 | |
| AK152467.1 | 4 | 426 | BAE31243.1 | |
| BC046820.1 | 4 | 412 | AAH46820.1 | |
| BC057196.1 | 4 | 331 | AAH57196.1 |

Genomic Map