Assay Details
Target Gene Details
Entrez Gene ID: | 76992 |
Gene Name: | family with sequence similarity 219, member A, opposite strand |
Gene Aliases: |
1700066J24Rik |
Location: |
Chr.4:41517437-41518929 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam219aos | NR_045726.1 | 1 | 163 | |
| AK006904.1 | 1 | 164 | BAB24786.1 |

Genomic Map